CAS RN 6381-79-9
CAS RN 6381-79-9
IUPAC Name:
tetrapotassium trihydrate dicarbonate
Synonyms:
tetrapotassium trihydrate dicarbonate
Molecular Weight (g/mol):
330.45
Molecular Formula:
C2H6K4O9
InChi Key:
WWVGGBSGYNQKCY-UHFFFAOYSA-J
SMILES:
O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O
1
–
4
of
4
results
1
Potassium carbonate sesquihydrate, ACS, 98.5-101.0%
CAS: 6381-79-9 Molecular Formula: C2H6K4O9 Molecular Weight (g/mol): 330.45 MDL Number: MFCD00149925 InChI Key: WWVGGBSGYNQKCY-UHFFFAOYSA-J IUPAC Name: tetrapotassium trihydrate dicarbonate SMILES: O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O
2
Potassium carbonate sesquihydrate, 98.5%, ACS reagent
CAS: 6381-79-9 Molecular Formula: C2H6K4O9 Molecular Weight (g/mol): 330.45 MDL Number: MFCD00149925 InChI Key: WWVGGBSGYNQKCY-UHFFFAOYSA-J IUPAC Name: tetrapotassium trihydrate dicarbonate SMILES: O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O