accessibility menu, dialog, popup

UserName

CAS RN 637-59-2

Br

CAS RN 637-59-2

IUPAC Name: (3-bromopropyl)benzene
Synonyms: propylbromide, G-phenyl
Molecular Weight (g/mol): 199.09
Molecular Formula: C9H11Br
InChi Key: XMZQWZJMTBCUFT-UHFFFAOYSA-N
SMILES: BrCCCC1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)