CAS RN 63286-28-2
CAS RN 63286-28-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloro-3-hydrazinylpyrazine, TRC
CAS: 63286-28-2 Molecular Formula: C4H5N4Cl Molecular Weight (g/mol): 144.56 IUPAC Name: (3-chloropyrazin-2-yl)hydrazine SMILES: NNc1nccnc1Cl