CAS RN 6324-10-3
IUPAC Name:
4,5-dibromothiophene-2-carboxylic acid
Synonyms:
4,5-dibromothiophene-2-carboxylic acid
Molecular Weight (g/mol):
285.94
Molecular Formula:
C5H2Br2O2S
InChi Key:
USFXRYVNRUMABJ-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(Br)=C(Br)S1
1
–
1
of
1
results
1
4,5-Dibromothiophene-2-carboxylic acid, 98%, Thermo Scientific™
CAS: 6324-10-3 MDL Number: MFCD00039682