accessibility menu, dialog, popup

UserName

CAS RN 6319-40-0

HOOBrNOO

CAS RN 6319-40-0

IUPAC Name: 4-bromo-3-nitrobenzoic acid
Synonyms: benzoic acid, 4-bromo-3-nitro-
Molecular Weight (g/mol): 246.02
Molecular Formula: C7H4BrNO4
InChi Key: RVCTZJVBWNFYRU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Br)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results