accessibility menu, dialog, popup

UserName

CAS RN 629672-19-1

CH3SOOClF

CAS RN 629672-19-1

IUPAC Name: 4-fluoro-3-methylbenzene-1-sulfonyl chloride
Synonyms: 4-fluoro-3-methylbenzenesulfonyl chloride
Molecular Weight (g/mol): 208.63
Molecular Formula: C7H6ClFO2S
InChi Key: ALEQALYEZFNWOP-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC(=C1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)