accessibility menu, dialog, popup

UserName

CAS RN 629-30-1

HOOH

CAS RN 629-30-1

IUPAC Name: heptane-1,7-diol
Synonyms: 1,7-heptanediol
Molecular Weight (g/mol): 132.2
Molecular Formula: C7H16O2
InChi Key: SXCBDZAEHILGLM-UHFFFAOYSA-N
SMILES: OCCCCCCCO
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)