CAS RN 629-25-4
CAS RN 629-25-4
IUPAC Name:
sodium dodecanoate
Synonyms:
sodium laurate
Molecular Weight (g/mol):
222.3
Molecular Formula:
C12H23NaO2
InChi Key:
BTURAGWYSMTVOW-UHFFFAOYSA-M
SMILES:
[Na+].CCCCCCCCCCCC([O-])=O
No Refinements
1
–
0
of
0
results
1
Lauric acid, sodium salt, 98%
CAS: 629-25-4 Molecular Formula: C12H23NaO2 Molecular Weight (g/mol): 222.30 MDL Number: MFCD00041754 InChI Key: BTURAGWYSMTVOW-UHFFFAOYSA-M Synonym: sodium dodecanoate,sodium laurate,lauric acid sodium salt,dodecanoic acid, sodium salt,lauric acid, sodium salt,dodecanoic acid sodium salt,lauran sodny czech,caswell no. 778a,unii-k146mr5exo,epa pesticide chemical code 079026 PubChem CID: 2735067 IUPAC Name: sodium;dodecanoate SMILES: [Na+].CCCCCCCCCCCC([O-])=O