CAS RN 628692-21-7
IUPAC Name:
2-(2-fluoro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms:
2-(2-fluoro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol):
252.09
Molecular Formula:
C13H18BFO3
InChi Key:
PYGFGOBGKWJERN-UHFFFAOYSA-N
SMILES:
COC1=CC(F)=C(C=C1)B1OC(C)(C)C(C)(C)O1
1
–
1
of
1
results
1
2-Fluoro-4-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 628692-21-7 Molecular Formula: C13H18BFO3 Molecular Weight (g/mol): 252.09 MDL Number: MFCD16996299 InChI Key: PYGFGOBGKWJERN-UHFFFAOYSA-N Synonym: 2-2-fluoro-4-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-fluoro-4-methoxybenzeneboronic acid pinacol ester,2-2-fluoro-4-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane,1,3,2-dioxaborolane, 2-2-fluoro-4-methoxyphenyl-4,4,5,5-tetramethyl PubChem CID: 17964843 IUPAC Name: 2-(2-fluoro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane SMILES: COC1=CC(F)=C(C=C1)B1OC(C)(C)C(C)(C)O1