accessibility menu, dialog, popup

UserName

CAS RN 62778-12-5

H3CNCH3NH2H3CNCH3NH2HOOOHO

CAS RN 62778-12-5

IUPAC Name: bis(N1,N1-dimethylbenzene-1,4-diamine); oxalic acid
Synonyms: bis(dimethyl-p-phenylenediamine); oxalic acid
Molecular Weight (g/mol): 362.43
Molecular Formula: C18H26N4O4
InChi Key: MNUINXKPLPIOEF-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.CN(C)C1=CC=C(N)C=C1.CN(C)C1=CC=C(N)C=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)