accessibility menu, dialog, popup

UserName

NHF

CAS RN 62778-11-4

IUPAC Name: pyridine hydrofluoride
Synonyms: pyridine hydrofluoride
Molecular Weight (g/mol): 99.11
Molecular Formula: C5H6FN
InChi Key: GRJJQCWNZGRKAU-UHFFFAOYSA-N
SMILES: F.C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results