accessibility menu, dialog, popup

UserName

CAS RN 627-86-1

H3COOOOCH3

CAS RN 627-86-1

IUPAC Name: 2-(octanoyloxy)ethyl octanoate
Synonyms: 2-(octanoyloxy)ethyl octanoate
Molecular Weight (g/mol): 314.47
Molecular Formula: C18H34O4
InChi Key: HTNFLUQQANUSLR-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)OCCOC(=O)CCCCCCC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results