accessibility menu, dialog, popup

UserName

CH3OHC

CAS RN 627-41-8

IUPAC Name: 3-methoxyprop-1-yne
Synonyms: 3-methoxy-propyne
Molecular Weight (g/mol): 70.09
Molecular Formula: C4H6O
InChi Key: YACFFSVYSPMSGS-UHFFFAOYSA-N
SMILES: COCC#C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)