CAS RN 6261-32-1
IUPAC Name:
(2Z)-2-(phenylmethylidene)-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms:
(2Z)-2-(phenylmethylidene)-3,4-dihydronaphthalen-1-one
Molecular Weight (g/mol):
234.3
Molecular Formula:
C17H14O
InChi Key:
JNRAEZULKWNOQO-QINSGFPZSA-N
SMILES:
O=C1\C(CCC2=CC=CC=C12)=C/C1=CC=CC=C1
1
–
1
of
1
results
1
2-Benzylidene-1-tetralone, 97%, Thermo Scientific™
CAS: 6261-32-1 Molecular Formula: C17H14O Molecular Weight (g/mol): 234.30 MDL Number: MFCD00019675,MFCD00019675 InChI Key: JNRAEZULKWNOQO-QINSGFPZSA-N Synonym: 2-benzylidene-1-tetralone,2z-2-benzylidenetetralin-1-one,2-benzylidenetetralone,2z-2-phenylmethylidene-3,4-dihydronaphthalen-1-one,1 2h-naphthalenone, 3,4-dihydro-2-phenylmethylene,2-benzylidene-1,2,3,4-tetrahydronaphthalen-1-one,2-benzylidene-3,4-dihydro-1 2h-naphthalenone,2z-2-benzylidene-3,4-dihydronaphthalen-1-one,2z-2-phenylmethylene-3,4-dihydronaphthalen-1-one PubChem CID: 6437615 IUPAC Name: (2Z)-2-benzylidene-3,4-dihydronaphthalen-1-one SMILES: O=C1\C(CCC2=CC=CC=C12)=C/C1=CC=CC=C1