accessibility menu, dialog, popup

UserName

CAS RN 626-48-2

CH3ONHOHN

CAS RN 626-48-2

IUPAC Name: 6-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 24PYRIMIDINEDIONE6methyl
Molecular Weight (g/mol): 126.12
Molecular Formula: C5H6N2O2
InChi Key: SHVCSCWHWMSGTE-UHFFFAOYSA-N
SMILES: CC1=CC(=O)NC(=O)N1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)