accessibility menu, dialog, popup

UserName

NaNaOHAsOHONHNOSOOONHNAsOHOHOOSOOO(E)(E)

CAS RN 62337-00-2

IUPAC Name: disodium (3E,6E)-3,6-bis[2-(2-arsonophenyl)hydrazin-1-ylidene]-4,5-dioxo-3,4,5,6-tetrahydronaphthalene-2,7-disulfonate
Synonyms: disodium (3E,6E)-3,6-bis[2-(2-arsonophenyl)hydrazin-1-ylidene]-4,5-dioxonaphthalene-2,7-disulfonate
Molecular Weight (g/mol): 820.33
Molecular Formula: C22H16As2N4Na2O14S2
InChi Key: QMDZRDUHVWMVAV-SQFCZRFJSA-L
SMILES: [Na+].[Na+].O[As](O)(=O)C1=CC=CC=C1N\N=C1/C(=O)C2=C(C=C1S([O-])(=O)=O)C=C(\C(=N\NC1=CC=CC=C1[As](O)(O)=O)C2=O)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results