accessibility menu, dialog, popup

UserName

CAS RN 62325-30-8

BrBr

CAS RN 62325-30-8

IUPAC Name: 2,7-dibromophenanthrene
Synonyms: 2,7-dibromophenanthrene
Molecular Weight (g/mol): 336.03
Molecular Formula: C14H8Br2
InChi Key: NGDWMVTZPZDKPM-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C(C=CC3=CC(Br)=CC=C23)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results