accessibility menu, dialog, popup

UserName

CAS RN 622-52-6

CH3NHNH2S

CAS RN 622-52-6

IUPAC Name: (4-methylphenyl)thiourea
Synonyms: urea, 2-thio-1-p-tolyl-
Molecular Weight (g/mol): 166.24
Molecular Formula: C8H10N2S
InChi Key: VXLFMCZPFIKKDZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC(N)=S)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results