CAS RN 62020-54-6
CAS RN 62020-54-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thiomuscimol, TRC
CAS: 62020-54-6 Molecular Formula: C4 H6 N2 O S Molecular Weight (g/mol): 130.17 IUPAC Name: 5-(aminomethyl)-1,2-thiazol-3-one SMILES: NCC1=CC(=O)NS1