accessibility menu, dialog, popup

UserName

CAS RN 620-24-6

HOHO

CAS RN 620-24-6

IUPAC Name: 3-(hydroxymethyl)phenol
Synonyms: 3-hydroxybenzyl alcohol M-hydroxybenzyl alcohol
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: OKVJCVWFVRATSG-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(O)=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (4)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (4)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (3)