accessibility menu, dialog, popup

UserName

CAS RN 616880-14-9

CH3H3COBOH3CH3CHNSCH3OO

CAS RN 616880-14-9

IUPAC Name: N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanesulfonamide
Synonyms: N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanesulfonamide
Molecular Weight (g/mol): 297.18
Molecular Formula: C13H20BNO4S
InChi Key: NPSPNWPZDXQYAC-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(NS(C)(=O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results