accessibility menu, dialog, popup

UserName

SCN

CAS RN 61499-10-3

IUPAC Name: (4-isothiocyanatobutyl)benzene
Synonyms: 4-phenylbutylisothiocyanate
Molecular Weight (g/mol): 191.29
Molecular Formula: C11H13NS
InChi Key: CCBQOLFAKKAMLD-UHFFFAOYSA-N
SMILES: S=C=NCCCCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results