CAS RN 61229-03-6
CAS RN 61229-03-6
IUPAC Name:
2-pentyloxirane
Synonyms:
2-pentyloxirane
Molecular Weight (g/mol):
114.19
Molecular Formula:
C7H14O
InChi Key:
NMOFYYYCFRVWBK-UHFFFAOYNA-N
SMILES:
CCCCCC1CO1