accessibility menu, dialog, popup

UserName

SOOHNOOHFFF

CAS RN 612042-13-4

IUPAC Name: 3-[3-(trifluoromethyl)benzenesulfonamido]propanoic acid
Synonyms: 3-[3-(trifluoromethyl)benzenesulfonamido]propanoic acid
Molecular Weight (g/mol): 297.25
Molecular Formula: C10H10F3NO4S
InChi Key: XNOHFTISYMPZGD-UHFFFAOYSA-N
SMILES: OC(=O)CCNS(=O)(=O)C1=CC=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results