accessibility menu, dialog, popup

UserName

CAS RN 612-62-4

ClN

CAS RN 612-62-4

IUPAC Name: 2-chloroquinoline
Synonyms: 2-chloroquinoline
Molecular Weight (g/mol): 163.6
Molecular Formula: C9H6ClN
InChi Key: OFUFXTHGZWIDDB-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C=CC=CC2=N1
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (4)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)