CAS RN 612-12-4
CAS RN 612-12-4
IUPAC Name:
1,2-bis(chloromethyl)benzene
Synonyms:
xylylene dichloride
Molecular Weight (g/mol):
175.05
Molecular Formula:
C8H8Cl2
InChi Key:
FMGGHNGKHRCJLL-UHFFFAOYSA-N
SMILES:
ClCC1=CC=CC=C1CCl
1
–
2
of
2
results
1
o-Xylylene dichloride, 98%
CAS: 612-12-4 Molecular Formula: C8H8Cl2 Molecular Weight (g/mol): 175.05 MDL Number: MFCD00000903 InChI Key: FMGGHNGKHRCJLL-UHFFFAOYSA-N Synonym: 1,2-bis chloromethyl benzene,o-xylylene dichloride,alpha,alpha'-dichloro-o-xylene,o-bis chloromethyl benzene,benzene, 1,2-bis chloromethyl,ccris 1773,o-xylene, alpha,alpha'-dichloro,benzene, 1,2-dichloromethyl,.alpha.,.alpha.'-dichloro-o-xylene,o-xylylenchlorid PubChem CID: 11919 IUPAC Name: 1,2-bis(chloromethyl)benzene SMILES: ClCC1=CC=CC=C1CCl