accessibility menu, dialog, popup

UserName

CAS RN 611-09-6

ONOHNOO

CAS RN 611-09-6

IUPAC Name: 5-nitro-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 1H-indole-2,3-dione, 5-nitro-
Molecular Weight (g/mol): 192.13
Molecular Formula: C8H4N2O4
InChi Key: UNMYHYODJHKLOC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2NC(=O)C(=O)C2=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)