accessibility menu, dialog, popup

UserName

CAS RN 610274-01-6

CH3SNNH3CCl

CAS RN 610274-01-6

IUPAC Name: 3-chloro-5,11-dimethyl-8-thia-4,6-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2,4,6-tetraene
Synonyms: 3-chloro-5,11-dimethyl-8-thia-4,6-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2,4,6-tetraene
Molecular Weight (g/mol): 252.76
Molecular Formula: C12H13ClN2S
InChi Key: DNHPYCAMLHXYLU-UHFFFAOYNA-N
SMILES: CC1CCC2=C(C1)SC1=NC(C)=NC(Cl)=C21
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results