accessibility menu, dialog, popup

UserName

CAS RN 606-21-3

ONOClNOO

CAS RN 606-21-3

IUPAC Name: 2-chloro-1,3-dinitrobenzene
Synonyms: benzene, 2-chloro-1,3-dinitro-
Molecular Weight (g/mol): 202.55
Molecular Formula: C6H3ClN2O4
InChi Key: BPPMIQPXQVIZNJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1Cl)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results