accessibility menu, dialog, popup

UserName

CH3OHOH3C

CAS RN 603-79-2

IUPAC Name: 2,3-dimethylbenzoic acid
Synonyms: 2,3-dimethylbenzoic acid
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: RIZUCYSQUWMQLX-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C(O)=O)=C1C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results