accessibility menu, dialog, popup

UserName

CAS RN 60177-39-1

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 344.95
Molecular Formula: C22H31ClN
InChi Key: —
SMILES: [Cl-].*-C=C.*-C=C.*-C=C.C1=CC=CC=C1.C[N+](C)(C)CC1=CC=CC=C1
Color 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)