accessibility menu, dialog, popup

UserName

CAS RN 6008-36-2

H3CO

CAS RN 6008-36-2

IUPAC Name: 1-phenylnonan-1-one
Synonyms: 1-phenylnonan-1-one
Molecular Weight (g/mol): 218.34
Molecular Formula: C15H22O
InChi Key: PFUPABFCHVRLLY-UHFFFAOYSA-N
SMILES: CCCCCCCCC(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results