accessibility menu, dialog, popup

UserName

CAS RN 6000-44-8

NaH2NOO

CAS RN 6000-44-8

IUPAC Name: sodium 2-aminoacetate
Synonyms: sodium glycinate
Molecular Weight (g/mol): 97.05
Molecular Formula: C2H4NNaO2
InChi Key: WUWHFEHKUQVYLF-UHFFFAOYSA-M
SMILES: [Na+].NCC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)