accessibility menu, dialog, popup

UserName

CAS RN 59878-57-8

IUPAC Name: 4-cyclopropanecarbonylpiperazin-1-ium
Synonyms: 4-cyclopropanecarbonylpiperazin-1-ium
Molecular Weight (g/mol): 155.22
Molecular Formula: C8H15N2O
InChi Key: KIALFUYSJAAJSU-UHFFFAOYSA-O
SMILES: O=C(C1CC1)N1CC[NH2+]CC1
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results