accessibility menu, dialog, popup

UserName

CH3H3CCH3OOOBr

CAS RN 59862-90-7

IUPAC Name: tert-butyl 3-bromofuran-2-carboxylate
Synonyms: tert-butyl 3-bromofuran-2-carboxylate
Molecular Weight (g/mol): 247.09
Molecular Formula: C9H11BrO3
InChi Key: STWBDYGNHJTHOY-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C1=C(Br)C=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results