accessibility menu, dialog, popup

UserName

CAS RN 5985-24-0

CH3OOOHOOH3C

CAS RN 5985-24-0

IUPAC Name: 1,3-dimethyl 4-hydroxybenzene-1,3-dicarboxylate
Synonyms: 1,3-dimethyl 4-hydroxybenzene-1,3-dicarboxylate
Molecular Weight (g/mol): 210.19
Molecular Formula: C10H10O5
InChi Key: ALBUJVBOIXVVLS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(O)C(=C1)C(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results