accessibility menu, dialog, popup

UserName

CAS RN 598-32-3

CH3HOH2C

CAS RN 598-32-3

IUPAC Name: but-3-en-2-ol
Synonyms: 3-buten-2-ol
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: MKUWVMRNQOOSAT-UHFFFAOYNA-N
SMILES: CC(O)C=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)