accessibility menu, dialog, popup

UserName

CH3OPOHOOH3C

CAS RN 598-02-7

IUPAC Name: diethoxyphosphinic acid
Synonyms: diethyl phosphoric acid diethyl acid phosphate (eto)2(HO)PO ethyl phosphate diethylphosphoric acid ethyl phosphate, (eto)2(HO)PO
Molecular Weight (g/mol): 154.1
Molecular Formula: C4H11O4P
InChi Key: UCQFCFPECQILOL-UHFFFAOYSA-N
SMILES: CCOP(O)(=O)OCC
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)