accessibility menu, dialog, popup

UserName

CH3SiOHH3CH3C

CAS RN 597-52-4

IUPAC Name: triethylsilanol
Synonyms: triethyl hydroxy silicon
Molecular Weight (g/mol): 132.28
Molecular Formula: C6H16OSi
InChi Key: WVMSIBFANXCZKT-UHFFFAOYSA-N
SMILES: CC[Si](O)(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results