accessibility menu, dialog, popup

UserName

NaNaNaNaOSOOSOOOSOOOSOOO

CAS RN 59572-10-0

IUPAC Name: tetrasodium pyrene-1,3,6,8-tetrasulfonate
Synonyms: tetrasodium pyrene-1,3,6,8-tetrasulfonate
Molecular Weight (g/mol): 610.41
Molecular Formula: C16H6Na4O12S4
InChi Key: UZBIRLJMURQVMX-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC(=C2C=CC3=C(C=C(C4=CC=C1C2=C34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)