accessibility menu, dialog, popup

UserName

CAS RN 594-09-2

CH3PCH3H3C

CAS RN 594-09-2

IUPAC Name: trimethylphosphane
Synonyms: trimethylphosphine Me3P trimethyl phosphine PMe3
Molecular Weight (g/mol): 76.08
Molecular Formula: C3H9P
InChi Key: YWWDBCBWQNCYNR-UHFFFAOYSA-N
SMILES: CP(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results