accessibility menu, dialog, popup

UserName

CAS RN 5933-40-4

H3CNHCH3

CAS RN 5933-40-4

IUPAC Name: methyl(1-phenylethyl)amine
Synonyms: methyl(1-phenylethyl)amine
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: RCSSHZGQHHEHPZ-UHFFFAOYNA-N
SMILES: CNC(C)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results