accessibility menu, dialog, popup

UserName

NOHONOO

CAS RN 59290-82-3

IUPAC Name: 3-nitropyridine-4-carboxylic acid
Synonyms: 3-nitropyridine-4-carboxylic acid
Molecular Weight (g/mol): 168.11
Molecular Formula: C6H4N2O4
InChi Key: YKPIUHZVTZWEEW-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=NC=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results