accessibility menu, dialog, popup

UserName

Cl

CAS RN 5911-08-0

IUPAC Name: (chloromethyl)cyclopropane
Synonyms: chloromethyl-cyclopropane
Molecular Weight (g/mol): 90.55
Molecular Formula: C4H7Cl
InChi Key: ZVTQWXCKQTUVPY-UHFFFAOYSA-N
SMILES: ClCC1CC1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)