accessibility menu, dialog, popup

UserName

CAS RN 59084-16-1

H3CONClO

CAS RN 59084-16-1

IUPAC Name: 1-acetylpiperidine-4-carbonyl chloride
Synonyms: 1-acetylpiperidine-4-carbonyl chloride
Molecular Weight (g/mol): 189.64
Molecular Formula: C8H12ClNO2
InChi Key: OHCPVLJEAHBMEG-UHFFFAOYSA-N
SMILES: CC(=O)N1CCC(CC1)C(Cl)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)