accessibility menu, dialog, popup

UserName

CAS RN 59-31-4

OHN

CAS RN 59-31-4

IUPAC Name: 1,2-dihydroquinolin-2-one
Synonyms: 2-oxoquinoline α-quinolone o-aminocinnamic acid lactam
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: LISFMEBWQUVKPJ-UHFFFAOYSA-N
SMILES: O=C1NC2=CC=CC=C2C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results