accessibility menu, dialog, popup

UserName

H3COCl

CAS RN 5891-21-4

IUPAC Name: 5-chloropentan-2-one
Synonyms: 5-chloropentan-2-one
Molecular Weight (g/mol): 120.58
Molecular Formula: C5H9ClO
InChi Key: XVRIEWDDMODMGA-UHFFFAOYSA-N
SMILES: CC(=O)CCCCl
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)