accessibility menu, dialog, popup

UserName

CAS RN 589-87-7

IUPAC Name: 1-bromo-4-iodobenzene
Synonyms: 4-bromoiodobenzene
Molecular Weight (g/mol): 282.91
Molecular Formula: C6H4BrI
InChi Key: UCCUXODGPMAHRL-UHFFFAOYSA-N
SMILES: BrC1=CC=C(I)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)