accessibility menu, dialog, popup

UserName

CAS RN 58577-88-1

OOHNH2

CAS RN 58577-88-1

IUPAC Name: 2-amino-3-(benzyloxy)propan-1-ol
Synonyms: 2-amino-3-(benzyloxy)propan-1-ol
Molecular Weight (g/mol): 181.24
Molecular Formula: C10H15NO2
InChi Key: ZJUOMDNENVWMPL-UHFFFAOYNA-N
SMILES: NC(CO)COCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results