accessibility menu, dialog, popup

UserName

CAS RN 58569-55-4

IUPAC Name: (2S)-2-[(2S)-2-(2-{2-[(2S)-2-amino-3-(4-hydroxyphenyl)propanamido]acetamido}acetamido)-3-phenylpropanamido]-4-(methylsulfanyl)butanoic acid
Synonyms: Met-enkephalin met-enkephalin
Molecular Weight (g/mol): 573.67
Molecular Formula: C27H35N5O7S
InChi Key: YFGBQHOOROIVKG-FKBYEOEOSA-N
SMILES: CSCC[C@H](NC(=O)[C@H](CC1=CC=CC=C1)NC(=O)CNC(=O)CNC(=O)[C@@H](N)CC1=CC=C(O)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)